missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11398606
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1623-92-3
Molecular Formula C12H9ClO3S
Molecular Weight (g/mol) 268.71
MDL Number MFCD00625748
InChI Key QIZPONOMFWAPRR-UHFFFAOYSA-N
Synonym 4-phenoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-phenoxy, 4-phenoxy-benzenesulfonyl chloride, 4-phenoxyphenylsulfonyl chloride, 4-phenoxybenzenesulphonyl chloride, 4-phenoxy benzene sulfonyl chloride, acmc-1bvtb, chloro 4-phenoxyphenyl sulfone, 4-phenoxybenzene-sulfonyl chloride, 4-phenoxy-benzenesulphonyl chloride
PubChem CID 2794756
IUPAC Name 4-phenoxybenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 42°C to 45°C
Boiling Point 180°C (5 mmHg)
Quantity 5 g
UN Number UN3260
Beilstein 2852659
Sensitivity Moisture sensitive
Formula Weight 268.72
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'