missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenoxybenzoic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.
Solubility
Very faint turbidity in hot methanol.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.
Solubility
Very faint turbidity in hot methanol.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 160°C to 164°C |
Quantity | 5 g |
Beilstein | 2212463 |
Solubility Information | Very faint turbidity in hot methanol. |
Formula Weight | 214.22 |
Percent Purity | 99% |
Chemical Name or Material | 4-Phenoxybenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'