missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-phenoxybenzoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | benzoyl chloride, 4-phenoxy, p-phenoxybenzoyl chloride, 4-phenoxybenzoylchloride, benzoyl chloride,4-phenoxy, p-phenoxybenzoylchloride, 4-phenoxy benzoyl chloride, 4-phenoxy-benzoyl chloride, 4-phenoxybenzoyl chlo-ride, benzoyl chloride, p-phenoxy, 4-phenoxyphenylcarbonyl chloride |
Formula Weight | 232.67 |
Percent Purity | 97% |
Chemical Name or Material | 4-Phenoxybenzoyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'