Learn More
4-phenoxybenzoyl chloride, 97%, Thermo Scientific™
£70.20 - £451.00
Chemical Identifiers
CAS | 1623-95-6 |
---|---|
Molecular Formula | C13H9ClO2 |
Molecular Weight (g/mol) | 232.663 |
InChI Key | AOOZVQGGMFGGEE-UHFFFAOYSA-N |
PubChem CID | 74193 |
IUPAC Name | 4-phenoxybenzoyl chloride |
SMILES | C1=CC=C(C=C1)OC2=CC=C(C=C2)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10516863
|
Thermo Scientific Maybridge
CC53702CB |
250 mg | Amber Glass Bottle |
£70.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10338063
|
Thermo Scientific Maybridge
CC53702DA |
1 g | Amber Glass Bottle |
£167.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10769225
|
Thermo Scientific Maybridge
CC53702DE |
5 g | Amber Glass Bottle |
£451.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1623-95-6 | |
232.663 | |
74193 | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(=O)Cl |
C13H9ClO2 | |
AOOZVQGGMFGGEE-UHFFFAOYSA-N | |
4-phenoxybenzoyl chloride |
Specifications
1623-95-6 | |
C13H9ClO2 | |
AOOZVQGGMFGGEE-UHFFFAOYSA-N | |
4-phenoxybenzoyl chloride | |
74193 | |
97% |
97% | |
benzoyl chloride, 4-phenoxy, p-phenoxybenzoyl chloride, 4-phenoxybenzoylchloride, benzoyl chloride,4-phenoxy, p-phenoxybenzoylchloride, 4-phenoxy benzoyl chloride, 4-phenoxy-benzoyl chloride, 4-phenoxybenzoyl chlo-ride, benzoyl chloride, p-phenoxy, 4-phenoxyphenylcarbonyl chloride | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(=O)Cl | |
232.663 | |
232.67 | |
4-Phenoxybenzoyl chloride |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.