Learn More
4-Phenoxyphenethylamine, 97+%, Thermo Scientific™
£58.50 - £142.00
Chemical Identifiers
CAS | 118468-18-1 |
---|---|
Molecular Formula | C14H15NO |
Molecular Weight (g/mol) | 213.28 |
MDL Number | MFCD00079777 |
InChI Key | JNHPLGDXCJAUBX-UHFFFAOYSA-N |
PubChem CID | 145535 |
IUPAC Name | 2-(4-phenoxyphenyl)ethanamine |
SMILES | C1=CC=C(C=C1)OC2=CC=C(C=C2)CCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10617274
|
Thermo Scientific Acros
393492500 |
250 mg | Glass Bottle |
£58.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10208383
|
Thermo Scientific Acros
393490010 |
1 g | Glass bottle | ||||||
Description
Chemical Identifiers
118468-18-1 | |
213.28 | |
JNHPLGDXCJAUBX-UHFFFAOYSA-N | |
2-(4-phenoxyphenyl)ethanamine |
C14H15NO | |
MFCD00079777 | |
145535 | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)CCN |
Specifications
118468-18-1 | |
100.0 | |
1.0900g/mL | |
Authentic | |
C14H15NO | |
1.5830 to 1.5850 (20°C, 589nm) | |
13, IV, 1789 | |
4-phenoxyphenethylamine, 2-4-phenoxyphenyl ethanamine, 2-4-phenoxyphenyl ethan-1-amine, o-phenyl-tyramine, benzeneethanamine,4-phenoxy, benzeneethanamine, 4-phenoxy, 2-4-phenoxy-phenyl-ethylamine, 4-butylpiperidine hcl form, acmc-1cb59, 2-4-phenoxyphenyl ethylamine | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)CCN | |
213.28 | |
213.28 | |
Liquid |
97.0 | |
Yellow | |
125°C (1.0 mmHg) | |
97% min. (GC) | |
C6H5OC6H4CH2CH2NH2 | |
MFCD00079777 | |
1.09 | |
JNHPLGDXCJAUBX-UHFFFAOYSA-N | |
2-(4-phenoxyphenyl)ethanamine | |
145535 | |
97+% | |
4-Phenoxyphenethylamine |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 4-Phenoxyphenethylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only