missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-buten-4-ol, 97%, Thermo Scientific Chemicals

Product Code. 10786183
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-58-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00039617
InChI Key RGKVZBXSJFAZRE-UHFFFAOYSA-N
Synonym 1-phenyl-3-buten-1-ol, 4-phenyl-1-buten-4-ol, 1-phenyl-but-3-en-1-ol, acmc-1bdtt, upenn_abs_025, 1-phenyl-3-buten-1-ol #, benzenemethanol,a-2-propen-1-yl, benzenemethanol, .alpha.-2-propenyl
PubChem CID 220119
IUPAC Name 1-phenylbut-3-en-1-ol
SMILES C=CCC(C1=CC=CC=C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9920g/mL
Boiling Point 234°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(OH)CH2CHCH2
Refractive Index 1.5300 to 1.532
Quantity 5 g
Specific Gravity 0.992
Formula Weight 148.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Phenyl-1-buten-4-ol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenyl-1-buten-4-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'