missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15471426
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1506428-22-3
Molecular Formula C14H9N3O2S
Molecular Weight (g/mol) 283.305
MDL Number MFCD20265306
InChI Key ZCNOYYRPSQWYSP-UHFFFAOYSA-N
Synonym 4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-phenyl-2-pyrazin-2-yl thiazole-5-carboxylic acid
PubChem CID 73995903
IUPAC Name 4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O

Specifications

Quantity 5 g
Formula Weight 283.3
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'