missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid, 97%, Thermo Scientific™

£40.69 - £588.00

Chemical Identifiers

CAS 1506428-22-3
Molecular Formula C14H9N3O2S
Molecular Weight (g/mol) 283.305
MDL Number MFCD20265306
InChI Key ZCNOYYRPSQWYSP-UHFFFAOYSA-N
Synonym 4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-phenyl-2-pyrazin-2-yl thiazole-5-carboxylic acid
PubChem CID 73995903
IUPAC Name 4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15481426
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54637.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15461426
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54637.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15471426
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54637.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1506428-22-3
283.305
ZCNOYYRPSQWYSP-UHFFFAOYSA-N
73995903
C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
C14H9N3O2S
MFCD20265306
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-phenyl-2-pyrazin-2-yl thiazole-5-carboxylic acid
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid

Specifications

1506428-22-3
MFCD20265306
ZCNOYYRPSQWYSP-UHFFFAOYSA-N
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
73995903
97%
C14H9N3O2S
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-phenyl-2-pyrazin-2-yl thiazole-5-carboxylic acid
C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
283.305
283.3
4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications