missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenylaminomethyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™
£40.59 - £136.00
Chemical Identifiers
CAS | 1029439-56-2 |
---|---|
Molecular Formula | C19H24BNO2 |
Molecular Weight (g/mol) | 309.216 |
MDL Number | MFCD06795652 |
InChI Key | KNYGNGFJIPTYBJ-UHFFFAOYSA-N |
Synonym | 4-n-phenylaminomethyl phenylboronic acid, pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl aniline, 4-n-phenylaminomethyl phenylboronic acid pinacol ester, 4-phenylaminomethyl benzeneboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl aniline, 4-phenylaminomethyl phenylboronic acid pinacol ester, phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzyl-amine, amtb054, 4-n-phenylaminomethyl phenylboronic acid |
PubChem CID | 17750276 |
IUPAC Name | N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]aniline |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CNC3=CC=CC=C3 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1029439-56-2 | |
309.216 | |
KNYGNGFJIPTYBJ-UHFFFAOYSA-N | |
17750276 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CNC3=CC=CC=C3 |
C19H24BNO2 | |
MFCD06795652 | |
4-n-phenylaminomethyl phenylboronic acid, pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl aniline, 4-n-phenylaminomethyl phenylboronic acid pinacol ester, 4-phenylaminomethyl benzeneboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl aniline, 4-phenylaminomethyl phenylboronic acid pinacol ester, phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzyl-amine, amtb054, 4-n-phenylaminomethyl phenylboronic acid | |
N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]aniline |
Specifications
1029439-56-2 | |
C19H24BNO2 | |
4-n-phenylaminomethyl phenylboronic acid, pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl aniline, 4-n-phenylaminomethyl phenylboronic acid pinacol ester, 4-phenylaminomethyl benzeneboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl aniline, 4-phenylaminomethyl phenylboronic acid pinacol ester, phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzyl-amine, amtb054, 4-n-phenylaminomethyl phenylboronic acid | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CNC3=CC=CC=C3 | |
309.216 | |
309.21 | |
4-(Phenylaminomethyl)benzeneboronic acid pinacol ester |
114°C to 117°C | |
MFCD06795652 | |
KNYGNGFJIPTYBJ-UHFFFAOYSA-N | |
N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]aniline | |
17750276 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures