missing translation for 'onlineSavingsMsg'
Learn More
4-phenylbutyl isothiocyanate, Thermo Scientific™ Product Code.: 10317422

4-phenylbutyl isothiocyanate, Thermo Scientific™

Product Code. 10317422
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 10317422

Brand: Thermo Scientific Maybridge TL00409DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61499-10-3
Molecular Formula C11H13NS
Molecular Weight (g/mol) 191.292
MDL Number MFCD00052335
InChI Key CCBQOLFAKKAMLD-UHFFFAOYSA-N
Synonym 4-phenylbutyl isothiocyanate, 4-phenylbutylisothiocyanate, 4-isothiocyanatobutyl benzene, phenylbutyl isothiocyanate, ccris 3148, 1-isothiocyanato-4-phenylbutane, 4-phenylbutylisolthiocyanate, benzene, 4-isothiocyanatobutyl, 4-phenylbutanisothiocyanate, 4-isothiocyanatobutyl benzene, 9ci
PubChem CID 124881
IUPAC Name 4-isothiocyanatobutylbenzene
SMILES C1=CC=C(C=C1)CCCCN=C=S

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 191.3
Percent Purity 95%
Chemical Name or Material 4-Phenylbutyl isothiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-phenylbutyl isothiocyanate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.