missing translation for 'onlineSavingsMsg'
Learn More
4-Phenylcarbamoylbenzeneboronic acid, 97%, Thermo Scientific™ Product Code.: 15489917

4-Phenylcarbamoylbenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15489917
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15489917

Brand: Thermo Scientific Alfa Aesar H53159.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330793-45-8
Molecular Formula C13H12BNO3
Molecular Weight (g/mol) 241.053
MDL Number MFCD04115683
InChI Key DAUSGSRIYCUJDK-UHFFFAOYSA-N
Synonym 4-phenylcarbamoyl phenyl boronic acid, 4-phenylcarbamoyl phenylboronic acid, 4-phenylaminocarbonylphenylboronic acid, 4-phenylaminocarbonylphenyl boronic acid, 4-phenylaminocarbonyl benzeneboronic acid, 4-phenylamino-1-carbonyl phenyl boronic acid, n-phenyl 4-boronobenzamide, acmc-1afql, 4-phenylcarbamoylbenzeneboronic acidShow MoreShow Less
PubChem CID 11096747
IUPAC Name [4-(phenylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2)(O)O

Specifications

Melting Point 210°C to 216°C
Quantity 250 mg
Formula Weight 241.05
Percent Purity 97%
Chemical Name or Material 4-Phenylcarbamoylbenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB-AH130301 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Phenylcarbamoylbenzeneboronic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.