Learn More
4-Phenylphenol, 97%, Thermo Scientific Chemicals
£40.20 - £151.00
Chemical Identifiers
CAS | 92-69-3 |
---|---|
Molecular Formula | C12H10O |
Molecular Weight (g/mol) | 170.21 |
MDL Number | MFCD00002347 |
InChI Key | YXVFYQXJAXKLAK-UHFFFAOYSA-N |
Synonym | 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol |
PubChem CID | 7103 |
ChEBI | CHEBI:34422 |
IUPAC Name | 4-phenylphenol |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011770
|
Thermo Scientific Acros
158712500 |
250 g | Plastic bottle |
£40.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686742
|
Thermo Scientific Acros
158710010 |
1 kg | Plastic bottle |
£151.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92-69-3 | |
170.21 | |
YXVFYQXJAXKLAK-UHFFFAOYSA-N | |
7103 | |
4-phenylphenol |
C12H10O | |
MFCD00002347 | |
4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol | |
CHEBI:34422 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)O |
Specifications
92-69-3 | |
100.0 | |
White to Yellow | |
165°C | |
96% min. (GC) | |
C6H5C6H4OH | |
06,674 | |
4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol | |
YXVFYQXJAXKLAK-UHFFFAOYSA-N | |
4-phenylphenol | |
7103 | |
170.21 | |
Flakes |
96.0 | |
>163.0°C | |
307.0°C | |
Authentic | |
C12H10O | |
MFCD00002347 | |
15,7416 | |
Solubility in water: 0.7g/L (20°C). Other solubilities: soluble in alkalies,soluble in most common organic solvents | |
C1=CC=C(C=C1)C2=CC=C(C=C2)O | |
170.21 | |
CHEBI:34422 | |
97% | |
4-Phenylphenol, 97% |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-179-2
missing translation for 'rtecsNumber' : DV5850000
missing translation for 'tsca' : TSCA
- 4-Phenylphenol
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only