missing translation for 'onlineSavingsMsg'
Learn More
4-Phenylpiperidine, 96%, Thermo Scientific Chemicals Product Code.: 15462739

4-Phenylpiperidine, 96%, Thermo Scientific Chemicals

Product Code. 15462739
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15462739

Brand: Thermo Scientific Alfa Aesar H66389.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-99-3
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00006002
InChI Key UTBULQCHEUWJNV-UHFFFAOYSA-N
Synonym 4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
PubChem CID 69873
IUPAC Name 4-phenylpiperidine
SMILES C1CNCCC1C2=CC=CC=C2

Specifications

Melting Point 61°C to 64°C
Color Yellow to Brown
Quantity 5 g
Beilstein 124508
Solubility Information Soluble in water.
Formula Weight 161.24
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Phenylpiperidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ29A048 Certificate TypeCertificate of Analysis Date25/07/2024 Catalog NumberH66389.03,H66389.06
Lot NumberQ29A048 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH66389.03,H66389.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Phenylpiperidine, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.