missing translation for 'onlineSavingsMsg'
Learn More

(4-phenyltetrahydropyran-4-yl)methylamine, 95%, Thermo Scientific™

Product Code. 10265474
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14006-32-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD02946795
InChI Key IPYDNTFWMDEOBW-UHFFFAOYSA-N
Synonym 4-phenyloxan-4-yl methanamine, 4-phenyltetrahydro-2h-pyran-4-yl methanamine, 4-phenyltetrahydropyran-4-yl methylamine, 1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, c-4-phenyl-tetrahydro-pyran-4-yl-methylamine, 4-phenyltetrahydro-2h-pyran-4-yl methylamine, 1-4-phenyloxan-4-yl methanamine, 2h-pyran-4-methanamine, tetrahydro-4-phenyl, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine, 4-phenyl-tetrahydro-2h-pyran-4-methylamine
PubChem CID 6483779
SMILES NCC1(CCOCC1)C1=CC=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-(4-phenyloxan-4-yl)methanamine
Formula Weight 191.27
Percent Purity 95%
Chemical Name or Material (4-phenyltetrahydropyran-4-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'