missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Picoline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
Solubility
Soluble in water, alcohol and ether.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
Solubility
Soluble in water, alcohol and ether.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 3°C |
Density | 0.957 |
pH | 10 to 11 |
Boiling Point | 144°C to 145°C |
Flash Point | 39°C (102°F) |
Odor | Unpleasant |
Refractive Index | 1.505 |
Quantity | 2500 mL |
UN Number | UN2313 |
Beilstein | 104586 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'