Learn More
4-Picoline, 98%, Thermo Scientific Chemicals
£19.50 - £89.30
Chemical Identifiers
CAS | 108-89-4 |
---|---|
Molecular Formula | C6H7N |
Molecular Weight (g/mol) | 93.129 |
MDL Number | MFCD00006440 |
InChI Key | FKNQCJSGGFJEIZ-UHFFFAOYSA-N |
Synonym | 4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy |
PubChem CID | 7963 |
ChEBI | CHEBI:32547 |
IUPAC Name | 4-methylpyridine |
SMILES | CC1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11477447
|
Thermo Scientific Alfa Aesar
A15595.AE |
100 mL |
£19.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487447
|
Thermo Scientific Alfa Aesar
A15595.AP |
500 mL |
£30.40
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497447
|
Thermo Scientific Alfa Aesar
A15595.0F |
2500 mL |
£89.30
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.
Solubility
Soluble in water, alcohol and ether.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
Chemical Identifiers
108-89-4 | |
93.129 | |
FKNQCJSGGFJEIZ-UHFFFAOYSA-N | |
7963 | |
4-methylpyridine |
C6H7N | |
MFCD00006440 | |
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy | |
CHEBI:32547 | |
CC1=CC=NC=C1 |
Specifications
108-89-4 | |
0.957 | |
144°C to 145°C | |
Unpleasant | |
1.505 | |
UN2313 | |
Air sensitive; Hygroscopic | |
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy | |
FKNQCJSGGFJEIZ-UHFFFAOYSA-N | |
4-methylpyridine | |
7963 | |
93.13 | |
4-Picoline |
3°C | |
10 to 11 | |
39°C (102°F) | |
C6H7N | |
MFCD00006440 | |
104586 | |
14,7402 | |
Soluble in water,alcohol and ether. | |
CC1=CC=NC=C1 | |
93.129 | |
CHEBI:32547 | |
98% |
Safety and Handling
GHS H Statement
H311-H226-H302-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H332-H311-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES
missing translation for 'einecsNumber' : 203-626-4
missing translation for 'rtecsNumber' : UT5425000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only