missing translation for 'onlineSavingsMsg'
Learn More

4-Piperazin-1-yl-furo[3,2-c]pyridine, ≥95%, Thermo Scientific™

Product Code. 10209882
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 81078-84-4
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD06411551
InChI Key LUIZVTKJWSGSPA-UHFFFAOYSA-N
Synonym 4-piperazin-1-yl furo 3,2-c pyridine, 4-piperazin-1-yl-furo 3,2-c pyridine, 1-furo 3,2-c pyridin-4-yl piperazine, furo 3,2-c pyridine, 4-1-piperazinyl, pubchem17312, d04vxp, 4-piperazinylfurano 3,2-c pyridine, 4-piperazin-1-ylfuro 3,2-c pyridine, 1-4-furo 3,2-c pyridinyl piperazine, 4-1-piperazinyl furo 3,2-c pyridine
PubChem CID 2794808
IUPAC Name 4-piperazin-1-ylfuro[3,2-c]pyridine
SMILES C1CN(CCN1)C2=NC=CC3=C2C=CO3

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material 4-Piperazin-1-yl-furo[3, 2-c]pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'