missing translation for 'onlineSavingsMsg'
Learn More

4-Piperazinoacetophenone, 94%, Thermo Scientific Chemicals

Product Code. 10532371
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51639-48-6
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.27
MDL Number MFCD00005962
InChI Key KPXVKKBJROCIJB-UHFFFAOYSA-N
Synonym 1-4-acetylphenyl piperazine, 1-4-piperazin-1-yl phenyl ethanone, 4'-piperazinoacetophenone, 1-4-piperazin-1-ylphenyl ethanone, 4-piperazinoacetophenone, piperazin-4-ylacetophenone, 1-4-piperazin-1-yl-phenyl-ethanone, p-piperazinoacetophenone, 1-4-piperazin-1-yl-phenyl ethanone
PubChem CID 97269
IUPAC Name 1-(4-piperazin-1-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2CCNCC2

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 107°C to 110°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 100 g
Formula Weight 204.27
Percent Purity 94%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Piperazinoacetophenone, 94%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Piperazinoacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.