missing translation for 'onlineSavingsMsg'
Learn More

4-(piperidin-1-ylmethyl)benzaldehyde, 95%, Thermo Scientific™

Product Code. 10598053
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 471929-86-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
InChI Key WWBOSCKXPMBVLG-UHFFFAOYSA-N
PubChem CID 7164649
IUPAC Name 4-(piperidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Synonym 4-piperidin-1-ylmethyl benzaldehyde, 4-1-piperidinylmethyl benzaldehyde, benzaldehyde,4-1-piperidinylmethyl, 4-piperidylmethyl benzaldehyde, 4-piperidinomethyl benzaldehyde, benzaldehyde, 4-1-piperidinylmethyl
Formula Weight 203.28
Percent Purity 95%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'