missing translation for 'onlineSavingsMsg'
Learn More
4-Piperidone ethylene ketal, 98+%, Thermo Scientific Chemicals Product Code.: 11429613

4-Piperidone ethylene ketal, 98+%, Thermo Scientific Chemicals

Product Code. 11429613
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11429613

Brand: Thermo Scientific Alfa Aesar A12419.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

Solubility
Soluble in water (Partly miscible).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177-11-7
Molecular Formula C7H14NO2
Molecular Weight (g/mol) 144.19
MDL Number MFCD00005976
InChI Key KPKNTUUIEVXMOH-UHFFFAOYSA-O
Synonym 1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
PubChem CID 67435
SMILES C1COC2(CC[NH2+]CC2)O1

Specifications

Density 1.117
Boiling Point 99°C to 101°C (14 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.4819
Quantity 250 g
Beilstein 506799
Sensitivity Air Sensitive
Solubility Information Soluble in water (Partly miscible).
IUPAC Name 1,4-dioxa-8-azaspiro[4.5]decane
Formula Weight 143.19
Percent Purity ≥98%
Chemical Name or Material 4-Piperidone ethylene ketal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Piperidone ethylene ketal, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.