missing translation for 'onlineSavingsMsg'
Learn More

4-PPBP maleate, Tocris Bioscience™

Product Code. 18033912
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specific σ ligand, NMDA antagonist

σ ligand and selective non-competitive antagonist at recombinant NR1a/2B NMDA receptors expressed in Xenopus oocytes.

Chemical Identifiers

CAS 201216-39-9
Molecular Formula C25H31NO4
Molecular Weight (g/mol) 409.526
InChI Key OASPNIMFGJVLES-WLHGVMLRSA-N
Synonym 4-ppbp maleate, 4-phenyl-1-4-phenylbutyl piperidine maleate salt
PubChem CID 71300190
IUPAC Name (E)-but-2-enedioic acid;4-phenyl-1-(4-phenylbutyl)piperidine
SMILES C1CN(CCC1C2=CC=CC=C2)CCCCC3=CC=CC=C3.C(=CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 409.52
Percent Purity >99%
Chemical Name or Material 4-PPBP maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.