missing translation for 'onlineSavingsMsg'
Learn More

(4-Pyrimidin-5-ylphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. 10413793
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 198084-13-8
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD06802874
InChI Key QPOIDCHBWLGFGU-UHFFFAOYSA-N
Synonym 4-pyrimidin-5-ylphenyl methanol, 4-pyrimidin-5-yl phenyl methanol, 4-5-pyrimidinyl phenyl methanol, 4-pyrimidin-5-yl-phenyl-methanol, 4-5-pyrimidinyl-benzyl alcohol, benzenemethanol,4-5-pyrimidinyl, 4-pyrimidin-5-ylphenyl methan-1-ol, swx
PubChem CID 7162046
IUPAC Name (4-pyrimidin-5-ylphenyl)methanol
SMILES C1=CC(=CC=C1CO)C2=CN=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material (4-Pyrimidin-5-ylphenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'