missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10295934
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 159691-25-5
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
InChI Key DOEIPZUTRVMWAB-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
PubChem CID 1092976
IUPAC Name 4-(pyrrolidin-1-ylmethyl)benzoic acid
SMILES C1CCN(C1)CC2=CC=C(C=C2)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'