missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Pyrrolidin-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™
£29.95 - £565.00
Chemical Identifiers
CAS | 159691-25-5 |
---|---|
Molecular Formula | C12H15NO2 |
Molecular Weight (g/mol) | 205.257 |
InChI Key | DOEIPZUTRVMWAB-UHFFFAOYSA-N |
Synonym | 4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride |
PubChem CID | 1092976 |
IUPAC Name | 4-(pyrrolidin-1-ylmethyl)benzoic acid |
SMILES | C1CCN(C1)CC2=CC=C(C=C2)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10295934
|
Thermo Scientific Maybridge
CC55601CB |
250 mg |
£29.95
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10441694
|
Thermo Scientific Maybridge
CC55601DA |
1 g |
£75.95
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10205944
|
Thermo Scientific Maybridge
CC55601EA |
10 g |
£565.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
159691-25-5 | |
205.257 | |
4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride | |
4-(pyrrolidin-1-ylmethyl)benzoic acid |
C12H15NO2 | |
DOEIPZUTRVMWAB-UHFFFAOYSA-N | |
1092976 | |
C1CCN(C1)CC2=CC=C(C=C2)C(=O)O |
Specifications
159691-25-5 | |
C12H15NO2 | |
DOEIPZUTRVMWAB-UHFFFAOYSA-N | |
4-(pyrrolidin-1-ylmethyl)benzoic acid | |
1092976 | |
97% |
97% | |
4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride | |
C1CCN(C1)CC2=CC=C(C=C2)C(=O)O | |
205.257 | |
205.26 | |
4-(Pyrrolidin-1-ylmethyl)benzoic acid |