missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 309.79 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'