missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzaldehyde, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 0.9700g/mL |
Boiling Point | 249°C |
Flash Point | 101°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)3CC6H4CHO |
Show More |
Safety and Handling
Product Identifier
- 4-tert-Butylbenzaldehyde
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Respiratory sensitiser Category 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'