missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10593721
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1710-98-1
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000695
InChI Key WNLMYNASWOULQY-UHFFFAOYSA-N
PubChem CID 74372
IUPAC Name 4-tert-butylbenzoyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)C(Cl)=O

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 135°C (20.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CC6H4COCl
Refractive Index 1.5354 to 1.5374
Quantity 25 g
Specific Gravity 1
Synonym benzoyl chloride, 4-1,1-dimethylethyl, 4-tert-butylbenzoylchloride, p-t-butylbenzoyl chloride, p-tert-butylbenzoyl chloride, benzoyl chloride,4-1,1-dimethylethyl, 4-tert-butyl-benzoyl chloride, 4-tert-butyl benzoyl chloride, 4-t-butylbenzoyl chloride, acmc-1brh5, 4-tertbutylbenzoyl chloride
Solubility Information Solubility in water: readily hydrolysed
Formula Weight 196.68
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzoyl chloride, 98%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 4-tert-Butylbenzoyl chloride
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'