missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl bromide, 97%, Thermo Scientific Chemicals

Product Code. 11421630
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18880-00-7
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00000180
InChI Key QZNQSIHCDAGZIA-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide
PubChem CID 87836
IUPAC Name 1-(bromomethyl)-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)CBr

Specifications

Melting Point 8°C to 12°C
Density 1.236
Boiling Point 93°C to 94°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5465
Quantity 5 g
UN Number UN3265
Beilstein 471674
Formula Weight 227.14
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'