missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals

£13.90 - £42.40

Chemical Identifiers

CAS 98-52-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00001473,MFCD00064952,MFCD00070476
InChI Key CCOQPGVQAWPUPE-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
PubChem CID 7391
IUPAC Name 4-tert-butylcyclohexan-1-ol
SMILES CC(C)(C)C1CCC(O)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10090580
View Documents
Thermo Scientific Acros
107900050
5 g Glass Bottle
£13.90
5g
Please to purchase this item. Need a web account? Register with us today!
10409210
View Documents
Thermo Scientific Acros
107902500
250 g Plastic bottle
£42.40
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-52-2
156.27
CCOQPGVQAWPUPE-UHFFFAOYSA-N
7391
CC(C)(C)C1CCC(O)CC1
C10H20O
MFCD00001473,MFCD00064952,MFCD00070476
4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
4-tert-butylcyclohexan-1-ol

Specifications

98-52-2
100.0
White
200°C
98.5% min. (GC)
(CH3)3CC6H10OH
MFCD00001473,MFCD00064952,MFCD00070476
4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
CCOQPGVQAWPUPE-UHFFFAOYSA-N
4-tert-butylcyclohexan-1-ol
7391
156.27
Powder or Granules
98.5
60.0°C to 69.0°C
224.0°C
Authentic
C10H20O
1.447
06, I, 18
Solubility in water: <1g/L water (20 c)
CC(C)(C)C1CCC(O)CC1
156.27
8.68 mPa.s (80°C)
99%
4-tert-Butylcyclohexanol, 99%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 202-676-4

missing translation for 'rtecsNumber' : GV8750000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only