missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetic acid, 98%

Product Code. 11408876
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11408876

Brand: Thermo Scientific Alfa Aesar A17017.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1798-04-5 | C12H16O3 | 208.257 g/mol

Applied as a reagent for the TAC protective group in nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied as a reagent for the TAC protective group in nucleosides.

Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1798-04-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00021758
InChI Key FBIGAJNVRFKBJL-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy
PubChem CID 15718
IUPAC Name 2-(4-tert-butylphenoxy)acetic acid
SMILES CC(C)(C)C1=CC=C(C=C1)OCC(=O)O

Specifications

Quantity 25 g
Solubility Information Sparingly Soluble (0.56g/L) (25°C),Calc.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 4-tert-Butylphenoxyacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.