missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butylsulfamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15419697
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208516-15-8
Molecular Formula C10H16BNO4S
Molecular Weight (g/mol) 257.11
MDL Number MFCD06659841
InChI Key BBTSLOMTYZIGGC-UHFFFAOYSA-N
Synonym 4-n-tert-butyl sulfamoyl phenyl boronic acid, 4-tert-butylaminosulphonyl benzeneboronic acid, t-butyl 4-boronobenzenesulfonamide, 4-tert-butylsulfamoyl phenylboronic acid, tert-butyl 4-boronobenzenesulfonamide, acmc-1ch0t, t-butyl 4-boronobenzene sulfonamide, 4-t-butylaminosulfonyl-phenylboronic acid, 4-tert-butylsulfamoyl-benzeneboronic acid, t-butyl 4-boronbenzenesulfonamide
PubChem CID 22107170
IUPAC Name [4-(tert-butylsulfamoyl)phenyl]boronic acid
SMILES CC(C)(C)NS(=O)(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 128°C to 132°C
Quantity 1 g
Formula Weight 257.11
Percent Purity 97%
Chemical Name or Material 4-(tert-Butylsulfamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'