missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)benzoyl chloride, Thermo Scientific™

Product Code. 10577484
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1151512-19-4
Molecular Formula C12H9ClOS
Molecular Weight (g/mol) 236.71
InChI Key AOHWXUBTRWZCOZ-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl benzoyl chloride, 4-2-thienylmethyl benzoyl chloride, 4-thiophen-2-ylmethyl benzoyl chloride, 4-thiophen-2-yl methyl benzoyl chloride
PubChem CID 45594296

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 236.72
Percent Purity 97%
Chemical Name or Material 4-(thien-2-ylmethyl)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'