missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(thien-2-ylmethyl)benzoyl chloride, Thermo Scientific™
£58.75 - £730.00
Chemical Identifiers
CAS | 1151512-19-4 |
---|---|
Molecular Formula | C12H9ClOS |
Molecular Weight (g/mol) | 236.71 |
InChI Key | AOHWXUBTRWZCOZ-UHFFFAOYSA-N |
Synonym | 4-thien-2-ylmethyl benzoyl chloride, 4-2-thienylmethyl benzoyl chloride, 4-thiophen-2-ylmethyl benzoyl chloride, 4-thiophen-2-yl methyl benzoyl chloride |
PubChem CID | 45594296 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10587864
|
Thermo Scientific Maybridge
CC70602CB |
250 mg | Amber Glass Bottle |
£58.75
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10577484
|
Thermo Scientific Maybridge
CC70602DA |
1 g | Amber Glass Bottle |
£182.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103864
|
Thermo Scientific Maybridge
CC70602DE |
5 g | Amber Glass Bottle |
£730.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1151512-19-4 | |
236.71 | |
4-thien-2-ylmethyl benzoyl chloride, 4-2-thienylmethyl benzoyl chloride, 4-thiophen-2-ylmethyl benzoyl chloride, 4-thiophen-2-yl methyl benzoyl chloride |
C12H9ClOS | |
AOHWXUBTRWZCOZ-UHFFFAOYSA-N | |
45594296 |
Specifications
1151512-19-4 | |
4-thien-2-ylmethyl benzoyl chloride, 4-2-thienylmethyl benzoyl chloride, 4-thiophen-2-ylmethyl benzoyl chloride, 4-thiophen-2-yl methyl benzoyl chloride | |
236.71 | |
236.72 | |
4-(thien-2-ylmethyl)benzoyl chloride |
C12H9ClOS | |
AOHWXUBTRWZCOZ-UHFFFAOYSA-N | |
45594296 | |
97% |
Safety and Handling
GHS Signal Word: Danger