missing translation for 'onlineSavingsMsg'
Learn More

4-Thien-2-yltetrahydropyran-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10693825
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-93-0
Molecular Formula C10H12O2S
Molecular Weight (g/mol) 196.264
MDL Number MFCD09817517
InChI Key BMUZFVLYGIZNAH-UHFFFAOYSA-N
Synonym 4-thiophen-2-yl oxane-4-carbaldehyde, 4-thien-2-yltetrahydropyran-4-carbaldehyde, 4-thien-2-yltetrahydro-2h-pyran-4-carboxaldehyde, 4-thiophen-2-yl tetrahydro-2h-pyran-4-carbaldehyde, 4-thiophen-2-yltetrahydropyran-4-carbaldehyde, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-carbaldehyde
PubChem CID 24229666
IUPAC Name 4-thiophen-2-yloxane-4-carbaldehyde
SMILES C1COCCC1(C=O)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material 4-Thien-2-yltetrahydropyran-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'