missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Thiouracil, 97%

Product Code. 10560754
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-28-6
Molecular Formula C4H4N2OS
Molecular Weight (g/mol) 128.15
MDL Number MFCD00090842
InChI Key OVONXEQGWXGFJD-UHFFFAOYSA-N
Synonym 4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci
PubChem CID 2734394
IUPAC Name 4-sulfanylidene-1H-pyrimidin-2-one
SMILES O=C1NC=CC(=S)N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 295°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 128.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Thiouracil, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Thiouracil
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'