missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethoxy)acetanilide, 98%, Thermo Scientific™

Product Code. 15474387
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1737-06-0
Melting Point 114°C to 117°C
Molecular Formula C9H8F3NO2
MDL Number MFCD00075293
Quantity 5 g
Synonym n-4-trifluoromethoxy phenyl acetamide, 4'-trifluoromethoxy acetanilide, 4-trifluoromethoxy acetanilide, 4-ocf3 acetanilide, acetamide, n-4-trifluoromethoxy phenyl, p-acetanisidide, alpha,alpha,alpha-trifluoro, n1-4-trifluoromethoxy phenyl acetamide, acetamide,n-4-trifluoromethoxy phenyl, maybridge1_000125
InChI Key QQRLAETWHFRNQH-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(OC(F)(F)F)C=C1
IUPAC Name N-[4-(trifluoromethoxy)phenyl]acetamide
Molecular Weight (g/mol) 219.16
PubChem CID 74437
Formula Weight 219.16
Percent Purity 98%
Chemical Name or Material 4'-(Trifluoromethoxy)acetanilide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'