Learn More
4-(Trifluoromethoxy)phenol, 98%, Thermo Scientific™
£25.00 - £244.00
Chemical Identifiers
CAS | 828-27-3 |
---|---|
Molecular Formula | C7H5F3O2 |
Molecular Weight (g/mol) | 178.11 |
MDL Number | MFCD00040988 |
InChI Key | WDRJNKMAZMEYOF-UHFFFAOYSA-N |
Synonym | 4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol |
PubChem CID | 70015 |
IUPAC Name | 4-(trifluoromethoxy)phenol |
SMILES | OC1=CC=C(OC(F)(F)F)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450603
|
Thermo Scientific Alfa Aesar
A12695.03 |
1 g | N/A | ||||||
11460603
|
Thermo Scientific Alfa Aesar
A12695.06 |
5 g |
£85.53
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470603
|
Thermo Scientific Alfa Aesar
A12695.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
828-27-3 | |
178.11 | |
WDRJNKMAZMEYOF-UHFFFAOYSA-N | |
70015 | |
OC1=CC=C(OC(F)(F)F)C=C1 |
C7H5F3O2 | |
MFCD00040988 | |
4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol | |
4-(trifluoromethoxy)phenol |
Specifications
828-27-3 | |
1.375 | |
86°C (186°F) | |
1.447 | |
UN2810 | |
Hygroscopic | |
WDRJNKMAZMEYOF-UHFFFAOYSA-N | |
4-(trifluoromethoxy)phenol | |
70015 | |
98% |
17°C to 18°C | |
91°C to 92°C (25 mmHg) | |
C7H5F3O2 | |
MFCD00040988 | |
1945934 | |
4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol | |
OC1=CC=C(OC(F)(F)F)C=C1 | |
178.11 | |
178.11 | |
4-(Trifluoromethoxy)phenol |
Safety and Handling
GHS H Statement
H301-H318-H227-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H301-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 212-583-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures