missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals

£76.80 - £266.00

Chemical Identifiers

CAS 68755-42-0
Molecular Formula C10H7F3O
Molecular Weight (g/mol) 200.16
MDL Number MFCD07772121
InChI Key LJVBFMQEZSEGRL-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1-indanone, 4-trifluoromethyl-2,3-dihydro-1h-inden-1-one, 4-trifluoromethyl-2,3-dihydroinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-trifluoromethyl, 4-trifluoromethyl indan-1-one, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one, 2,3-dihydro-1-oxo-4-trifluoromethyl-1h-indene, acmc-1b8x3, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one
PubChem CID 21921093
IUPAC Name 4-(trifluoromethyl)-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415398
View Documents
Thermo Scientific Alfa Aesar
H59165.03
1 g
£76.80
1g
Please to purchase this item. Need a web account? Register with us today!
15425398
View Documents
Thermo Scientific Alfa Aesar
H59165.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68755-42-0
200.16
LJVBFMQEZSEGRL-UHFFFAOYSA-N
21921093
C1CC(=O)C2=C1C(=CC=C2)C(F)(F)F
C10H7F3O
MFCD07772121
4-trifluoromethyl-1-indanone, 4-trifluoromethyl-2,3-dihydro-1h-inden-1-one, 4-trifluoromethyl-2,3-dihydroinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-trifluoromethyl, 4-trifluoromethyl indan-1-one, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one, 2,3-dihydro-1-oxo-4-trifluoromethyl-1h-indene, acmc-1b8x3, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one
4-(trifluoromethyl)-2,3-dihydroinden-1-one

Specifications

68755-42-0
67°C to 72°C (0.1 mmHg)
C10H7F3O
4-trifluoromethyl-1-indanone, 4-trifluoromethyl-2,3-dihydro-1h-inden-1-one, 4-trifluoromethyl-2,3-dihydroinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-trifluoromethyl, 4-trifluoromethyl indan-1-one, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one, 2,3-dihydro-1-oxo-4-trifluoromethyl-1h-indene, acmc-1b8x3, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one
C1CC(=O)C2=C1C(=CC=C2)C(F)(F)F
200.16
200.16
4-(Trifluoromethyl)-1-indanone
38°C to 42°C
>108°C (226°F)
MFCD07772121
LJVBFMQEZSEGRL-UHFFFAOYSA-N
4-(trifluoromethyl)-2,3-dihydroinden-1-one
21921093
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only