missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)anisole, 98%, Thermo Scientific Chemicals

Product Code. 15451279
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-52-8
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD03094456
InChI Key CFIPQRIPCRRISV-UHFFFAOYSA-N
Synonym 4-trifluoromethyl anisole, 1-methoxy-4-trifluoromethyl benzene, 4-trifluoromethylanisole, benzene, 1-methoxy-4-trifluoromethyl, 4-methoxybenzotrifluoride, buttpark 180\07-83, p-trifluoromethylanisole, pubchem10126, 4-trifluoromethyl-anisol, 1-methoxy-4-trifluoromethyl-benzene
PubChem CID 600604
IUPAC Name 1-methoxy-4-(trifluoromethyl)benzene
SMILES COC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point -9°C
Color Colorless to Yellow
Density 1.245 g/mL
Boiling Point 169°C (754 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.445
Quantity 1 g
UN Number UN1993
Formula Weight 176.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)anisole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'