Learn More
4-(Trifluoromethyl)benzaldehyde, 97%, Thermo Scientific Chemicals
£49.14 - £148.74
Chemical Identifiers
CAS | 455-19-6 |
---|---|
Molecular Formula | C8H5F3O |
Molecular Weight (g/mol) | 174.122 |
MDL Number | MFCD00006952 |
InChI Key | BEOBZEOPTQQELP-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde |
PubChem CID | 67996 |
IUPAC Name | 4-(trifluoromethyl)benzaldehyde |
SMILES | C1=CC(=CC=C1C=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11442338
|
Thermo Scientific Alfa Aesar
A15276.06 |
5 g |
£49.14
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452338
|
Thermo Scientific Alfa Aesar
A15276.14 |
25 g |
£148.74
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.
Solubility
Soluble in water. 1.5 g/L at 20°C
Notes
Air Sensitive. Store away from air and oxidizing agents. Store under an inert atmosphere. Incompatible with oxidizing agents, reducing agents.
Chemical Identifiers
455-19-6 | |
174.122 | |
BEOBZEOPTQQELP-UHFFFAOYSA-N | |
67996 | |
C1=CC(=CC=C1C=O)C(F)(F)F |
C8H5F3O | |
MFCD00006952 | |
4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde | |
4-(trifluoromethyl)benzaldehyde |
Specifications
455-19-6 | |
1.275 | |
65°C (149°F) | |
1.463 | |
1101680 | |
4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde | |
BEOBZEOPTQQELP-UHFFFAOYSA-N | |
4-(trifluoromethyl)benzaldehyde | |
67996 | |
97% |
1°C to 2°C | |
80°C to 81°C (25 mmHg) | |
C8H5F3O | |
MFCD00006952 | |
Air Sensitive | |
Soluble in water. 1.5g/L at 20°C | |
C1=CC(=CC=C1C=O)C(F)(F)F | |
174.122 | |
174.12 | |
4-(Trifluoromethyl)benzaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H320-H335
missing translation for 'einecsNumber' : 207-240-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.