missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Codice prodotto. 10387734
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Questo articolo non è restituibile. Consulta la politica di reso
Questo articolo non è restituibile. Consulta la politica di reso

Identificatori chimici

CAS 2991-42-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.61
MDL Number MFCD00042422
InChI Key OZDCZHDOIBUGAJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
PubChem CID 2777399
IUPAC Name 4-(trifluoromethyl)benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifica

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CF3OC6H4SO2Cl
Quantity 10 g
Formula Weight 244.62
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'