missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Spécification
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CF3OC6H4SO2Cl |
Quantity | 25 g |
Formula Weight | 244.62 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Trifluoromethyl)benzene-1-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'