missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™
£19.60 - £38.00
Chemical Identifiers
CAS | 2991-42-6 |
---|---|
Molecular Formula | C7H4ClF3O2S |
Molecular Weight (g/mol) | 244.61 |
MDL Number | MFCD00042422 |
InChI Key | OZDCZHDOIBUGAJ-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl |
PubChem CID | 2777399 |
IUPAC Name | 4-(trifluoromethyl)benzenesulfonyl chloride |
SMILES | FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387734
|
Thermo Scientific Maybridge
TL00175EA |
10 g | Amber Glass Bottle |
£19.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10586395
|
Thermo Scientific Maybridge
TL00175EB |
25 g | Amber Glass Bottle |
£38.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
2991-42-6 | |
244.61 | |
OZDCZHDOIBUGAJ-UHFFFAOYSA-N | |
2777399 | |
FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O |
C7H4ClF3O2S | |
MFCD00042422 | |
4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl | |
4-(trifluoromethyl)benzenesulfonyl chloride |
Specifications
2991-42-6 | |
C7H4ClF3O2S | |
MFCD00042422 | |
OZDCZHDOIBUGAJ-UHFFFAOYSA-N | |
4-(trifluoromethyl)benzenesulfonyl chloride | |
2777399 | |
97% |
97% | |
CF3OC6H4SO2Cl | |
4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl | |
FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O | |
244.61 | |
244.62 | |
4-(Trifluoromethyl)benzene-1-sulfonyl chloride |