missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzeneboronic acid, 98%

Product Code. 11410477
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11410477

Brand: Thermo Scientific Alfa Aesar B22374.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128796-39-4 | C7H6BF3O2 | 189.93 g/mol

4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

Solubility
Soluble in DMSO, not in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128796-39-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151855
InChI Key ALMFIOZYDASRRC-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
PubChem CID 2734389
IUPAC Name [4-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 238°C to 242°C
Quantity 1 g
Beilstein 3544189
Solubility Information Soluble in DMSO,not in water
Formula Weight 189.93
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzeneboronic acid
compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)benzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-(Trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.