missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11452237
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-15-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000694
InChI Key OXZYBOLWRXENKT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
PubChem CID 67607
IUPAC Name 4-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point -3°C
Density 1.404
Boiling Point 188°C to 190°C
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.477
Quantity 100 g
UN Number UN3265
Beilstein 391282
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 208.57
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'