missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylhydrazine, 95%

Product Code. 11393156
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11393156

Brand: Thermo Scientific Alfa Aesar L12436.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 368-90-1 | C7H7F3N2 | 176.14 g/mol

4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-90-1
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.14
MDL Number MFCD00042508
InChI Key DBNLGTYGKCMLLR-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylhydrazine, 4-trifluoromethyl phenyl hydrazine, 4-trifluoromethylphenylhydrazine, p-trifluoromethylphenylhydrazine, hydrazine, 4-trifluoromethyl phenyl, p-trifluoromethyl phenylhydrazine, 4-trifluoromethyl-phenylhydrazine, 4-trifluoromethyl-phenyl-hydrazine
PubChem CID 136200
IUPAC Name [4-(trifluoromethyl)phenyl]hydrazine
SMILES NNC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 60°C to 64°C
Boiling Point 119°C to 121°C (17 mmHg)
Quantity 5 g
Beilstein 743759
Solubility Information Insoluble in water.
Formula Weight 176.14
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine
compliance-icons compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)phenylhydrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : 4-(Trifluoromethyl)phenylhydrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.