missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine hydrochloride, 97%

Product Code. 11308746
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 11308746

missing translation for 'mfr': Thermo Scientific Alfa Aesar L19441.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 155849-49-3 | C6H11ClF3N | 189.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155849-49-3
Molecular Formula C6H11ClF3N
Molecular Weight (g/mol) 189.606
MDL Number MFCD04971993
InChI Key KMUKXPQVPQUCDB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine hydrochloride, 4-trifluoromethyl piperidine hcl, 4-trifluoromethyl piperidinehydrochloride, 4-trifluoromethylpiperidine hydrochloride, 4-trifluoromethyl-piperidine hydrochloride, piperidine, 4-trifluoromethyl-, hydrochloride, 4-trifluoromethyl piperidine, chloride, pubchem10319, 4-trifluoromethyl piperidine, hcl, 4-trifluoromethylpiperidine-hcl salt
PubChem CID 16218160
IUPAC Name 4-(trifluoromethyl)piperidine;hydrochloride
SMILES C1CNCCC1C(F)(F)F.Cl

Specifications

Melting Point ≥160°C (decomposition)
Quantity 250 mg
Beilstein 1341222
Formula Weight 189.61
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)piperidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.