missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11398736
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155849-49-3
Molecular Formula C6H11ClF3N
Molecular Weight (g/mol) 189.606
MDL Number MFCD04971993
InChI Key KMUKXPQVPQUCDB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine hydrochloride, 4-trifluoromethyl piperidine hcl, 4-trifluoromethyl piperidinehydrochloride, 4-trifluoromethylpiperidine hydrochloride, 4-trifluoromethyl-piperidine hydrochloride, piperidine, 4-trifluoromethyl-, hydrochloride, 4-trifluoromethyl piperidine, chloride, pubchem10319, 4-trifluoromethyl piperidine, hcl, 4-trifluoromethylpiperidine-hcl salt
PubChem CID 16218160
IUPAC Name 4-(trifluoromethyl)piperidine;hydrochloride
SMILES C1CNCCC1C(F)(F)F.Cl

Specifications

Melting Point ≥160°C (decomposition)
Quantity 1 g
Beilstein 1341222
Formula Weight 189.61
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)piperidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'