missing translation for 'onlineSavingsMsg'
Learn More

4-(Trimethylsilyl)diphenylacetylene, 99%, Thermo Scientific Chemicals

Product Code. 11471150
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136459-72-8
Molecular Formula C17H18Si
Molecular Weight (g/mol) 250.416
MDL Number MFCD09953470
InChI Key UZRJWRFLYPENFH-UHFFFAOYSA-N
Synonym 4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene
PubChem CID 21093331
IUPAC Name trimethyl-[4-(2-phenylethynyl)phenyl]silane
SMILES C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2

Specifications

Melting Point 37°C to 39°C
Quantity 5 g
Formula Weight 250.42
Percent Purity 99%
Chemical Name or Material 4-(Trimethylsilyl)diphenylacetylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'