missing translation for 'onlineSavingsMsg'
Learn More

4-Tritylphenol, 98%, Thermo Scientific Chemicals

Product Code. 11352356
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 978-86-9
Molecular Formula C25H20O
Molecular Weight (g/mol) 336.43
MDL Number MFCD00002364
InChI Key NIPKXTKKYSKEON-UHFFFAOYSA-N
Synonym 4-triphenylmethylphenol, 4-triphenylmethyl phenol, p-tritylphenol, p-triphenylmethylphenol, phenol, 4-triphenylmethyl, p-cresol, .alpha.,.alpha.,.alpha.-triphenyl, 4-tritylphenol, p-triphenylmethyl phenol, 4-tri phenyl methyl phenol, phenol,4-triphenylmethyl
PubChem CID 70422
IUPAC Name 4-tritylphenol
SMILES OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 276°C to 286°C
Quantity 25 g
Beilstein 2290531
Formula Weight 336.43
Percent Purity 98%
Chemical Name or Material 4-Tritylphenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'